Skip to Content
Merck
All Photos(1)

Documents

799521

Sigma-Aldrich

Indole-15N

98 atom % 15N, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H715N
CAS Number:
Molecular Weight:
118.14
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % 15N

Assay

98% (CP)

form

solid

mass shift

M+1

SMILES string

C12=CC=CC=C1C=C[15N]2

InChI

1S/C8H6N/c1-2-4-8-7(3-1)5-6-9-8/h1-6H/i9+1

InChI key

DAIGRYNHHZOIRD-QBZHADDCSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service