779784
(R)-(+)-2,2′-Dimethoxy-1,1′-binaphthalene
99%
Synonym(s):
(R)-(+)-2,2′-Dimethoxy-1,1′-binaphthyl, (R)-1,1′-Bi-2-naphthyl dimethyl ether
About This Item
Recommended Products
Quality Level
Assay
≥98.5% (HPLC)
99%
form
powder
optical activity
[α]20/D +52°, c = 1% in chloroform
mp
227-231 °C (lit.)
InChI
1S/C22H18O2/c1-23-19-13-11-15-7-3-5-9-17(15)21(19)22-18-10-6-4-8-16(18)12-14-20(22)24-2/h3-14H,1-2H3
InChI key
BJAADAKPADTRCH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service