Skip to Content
Merck

765910

Potassium tri(3,5-dimethyl-1-pyrazolyl)borohydride

97%, solid

Synonym(s):

Potassium hydrotris(3,5-dimethylpyrazol-1-yl)borate, Scorpionates, Tp ligand

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H22BKN6
CAS Number:
Molecular Weight:
336.28
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22
MDL number:
Assay:
97%
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Potassium tri(3,5-dimethyl-1-pyrazolyl)borohydride, 97%

Quality Segment

assay

97%

form

solid

mp

292-301 °C

SMILES string

[K+].Cc1cc(C)n(n1)[BH-](n2nc(C)cc2C)n3nc(C)cc3C

InChI

1S/C15H22BN6.K/c1-10-7-13(4)20(17-10)16(21-14(5)8-11(2)18-21)22-15(6)9-12(3)19-22;/h7-9,16H,1-6H3;/q-1;+1

InChI key

NTWZGFNSHCFHIJ-UHFFFAOYSA-N

Application

Scorpionate ligand


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library