Skip to Content
Merck
All Photos(1)

Key Documents

69819

Sigma-Aldrich

(−)-Butyl L-lactate

≥97.0% (sum of enantiomers, GC)

Synonym(s):

Butyl (2S)-2-hydroxypropanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O3
CAS Number:
Molecular Weight:
146.18
Beilstein:
1721597
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D −13±1°, neat

bp

183-190 °C (lit.)

density

0.981 g/mL at 20 °C (lit.)

SMILES string

CCCCOC(=O)[C@H](C)O

InChI

1S/C7H14O3/c1-3-4-5-10-7(9)6(2)8/h6,8H,3-5H2,1-2H3/t6-/m0/s1

InChI key

MRABAEUHTLLEML-LURJTMIESA-N

Application

(−)-Butyl L-lactate can be used as a precursor to synthesize poly(lactic acid) and in the synthesis of chiral mesogens (liquid crystals). It can also be used as a component in various pesticide formulations.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lactic acid, n-?butyl ester, (S) and lactic acid, ethyl ester, (S)?; exemption from the requirement of a tolerance.
Federal Register, 69(18), 4073-4077 (2004)
The chiral pool as valuable natural source: new chiral mesogens made from lactic acid.
Brombach F, et al.
Mol. Cryst. Liq. Cryst., 542(1), 62-584 (2011)
Bulk polytransesterification of L?lactic acid esters: An alternative route to synthesize poly (lactic acid).
Marques D S, et al.
Journal of Applied Polymer Science, 125(S2) (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service