Skip to Content
Merck
All Photos(1)

Documents

604003

Sigma-Aldrich

Acetanilide-(ring-13C6, carbonyl-13C)

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH313CONH13C6H5
CAS Number:
Molecular Weight:
142.11
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

99 atom % 13C

form

solid

bp

304 °C (lit.)

mp

113-115 °C (lit.)

density

1.283 g/mL at 25 °C

mass shift

M+7

SMILES string

C[13C](=O)N[13c]1[13cH][13cH][13cH][13cH][13cH]1

InChI

1S/C8H9NO/c1-7(10)9-8-5-3-2-4-6-8/h2-6H,1H3,(H,9,10)/i2+1,3+1,4+1,5+1,6+1,7+1,8+1

InChI key

FZERHIULMFGESH-NWWRDLAKSA-N

General description

Acetanilide-13C7, isotopically labeled analogue of acetanilide.

Application

Acetanilide-13C7 is used to study interaction sites of acetanilide with organic matter surrogates using nuclear magnetic resonance (NMR) techniques.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

321.8 °F - closed cup

Flash Point(C)

161.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shalini Jayasundera et al.
Journal of agricultural and food chemistry, 51(13), 3829-3835 (2003-06-12)
The dynamics of acetanilide pesticide interactions with organic matter (OM) surrogates were examined using nuclear magnetic resonance (NMR) spectroscopy. Differences in the relative changes in (13)C and (1)H spin-lattice relaxation times (T(1)) were measured at multiple molecular sites of metolachlor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service