Skip to Content
Merck
All Photos(1)

Key Documents

578401

Sigma-Aldrich

3-Cyanophenylboronic acid pinacol ester

97%

Synonym(s):

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16BNO2
CAS Number:
Molecular Weight:
229.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

78-82 °C (lit.)

functional group

nitrile

SMILES string

CC1(C)OB(OC1(C)C)c2cccc(c2)C#N

InChI

1S/C13H16BNO2/c1-12(2)13(3,4)17-14(16-12)11-7-5-6-10(8-11)9-15/h5-8H,1-4H3

InChI key

FIGQEPXOSAFKTA-UHFFFAOYSA-N

Application

3-Cyanophenylboronic acid pinacol ester can be used as a reactant to synthesize:
  • 3-Cyano aryl/ heteroaryl derivatives by forming a C−C bond via palladium-catalyzed Suzuki-Miyaura reaction.
  • 3-(Phenylamino)benzonitrile by copper-catalyzed Chan-Evans-Lam amination reaction.
  • 3-(Trifluoromethyl)benzonitrile using potassium (trifluoromethyl)trimethoxyborate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chan-Evans-Lam amination of boronic acid pinacol (BPin) esters: overcoming the aryl amine problem
Vantourout JC, et al.
The Journal of Organic Chemistry, 81(9), 3942-3950 (2016)
Speciation control during Suzuki-Miyaura cross-coupling of haloaryl and haloalkenyl MIDA boronic esters
Fyfe JWB, et al.
Chemistry?A European Journal , 21(24), 8951-8964 (2015)
Oxidative Trifluoromethylation of Arylboronates with Shelf-Stable Potassium (Trifluoromethyl) trimethoxyborate
Khan BA, et al.
Chemistry?A European Journal , 18(6), 1577-1581 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service