Skip to Content
Merck
All Photos(3)

Documents

478687

Sigma-Aldrich

4-Iodophenyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4NCO
CAS Number:
Molecular Weight:
245.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

44-48 °C (lit.)

storage temp.

2-8°C

SMILES string

Ic1ccc(cc1)N=C=O

InChI

1S/C7H4INO/c8-6-1-3-7(4-2-6)9-5-10/h1-4H

InChI key

KVMKUTIUJGYHBA-UHFFFAOYSA-N

Application

4-Iodophenyl isocyanate may be used in the preparation of:
  • silicon-stereogenic silicon-bridged arylpyridinones
  • N-(4-iodophenyl)cholesteryl-3-carbamate
  • N-(4-iodophenyl)pregnenolone-3-carbamate

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Van Dort et al.
International journal of radiation applications and instrumentation. Part B, Nuclear medicine and biology, 16(6), 603-607 (1989-01-01)
A series of radioiodinated benzoate and carbamate esters of cholesterol and pregnenolone wherein the acyl moiety served as the carrier for radioiodine was synthesized and evaluated as potential imaging agents for the adrenal cortex. 2,6-Dimethyl-3-iodobenzoyl and N-(4-iodophenyl) carbamoyl groups were
Rhodium-catalyzed asymmetric synthesis of silicon-stereogenic silicon-bridged arylpyridinones.
Shintani R, et al.
Chemical Science, 7, 1205-1211 (2016)
Franky Ka-Wah Hau et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 22(11), 3738-3749 (2016-02-03)
A series of mononuclear and dinuclear platinum(II) terpyridine complexes with amide-, sulfonamide-, and urea-containing ligands (1-9) has been successfully designed and synthesized, and their photophysical and anion-binding properties have been studied. The anion-binding properties of the complexes were studied by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service