Skip to Content
Merck
All Photos(2)

Key Documents

433829

Sigma-Aldrich

4-Hydrazinobenzoic acid hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NNHC6H4CO2H · HCl
CAS Number:
Molecular Weight:
188.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

253 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

Cl.NNc1ccc(cc1)C(O)=O

InChI

1S/C7H8N2O2.ClH/c8-9-6-3-1-5(2-4-6)7(10)11;/h1-4,9H,8H2,(H,10,11);1H

InChI key

XHLQMKQBCHYRLC-UHFFFAOYSA-N

General description

4-Hydrazinobenzoic acid (p-Hydrazinobenzoic acid, HBA) has been reported as an ingredient of the cultivated mushroom Agaricus bisporus. It is carcinogenic to rodents. Mechanism of DNA damage by 4-hydrazinobenzoic acid has been studied by using 32P-labeled DNA fragments obtained from the human p53 and p16 tumor suppressor genes. It may be used for the preparation of 4-(3-(2-(tert-butoxycarbonylamino)ethyl)-5-hydroxy-1H-pyrazol-1-yl)benzoic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 3-(2-aminoethyl)-5-hydroxy-1H-pyrazole derivatives.
Groselj U, et al.
ARKIVOC (Gainesville, FL, United States), 3, 49-65 (2012)
Yan-Juan Gu et al.
International journal of nanomedicine, 6, 2889-2898 (2011-12-02)
Single-walled carbon nanotubes (SWNTs) have been identified as an efficient drug carrier. Here a controlled drug-delivery system based on SWNTs coated with doxorubicin (DOX) through hydrazone bonds was developed, because the hydrazone bond is more sensitive to tumor microenvironments than
B M McManus et al.
Laboratory investigation; a journal of technical methods and pathology, 57(1), 78-85 (1987-07-01)
p-Hydrazinobenzoic acid (HBA), an ingredient of the cultivated mushroom Agaricus bisporus, was given in hydrochloride form at a dosage of 0.125% in drinking water for life to randomly bred Swiss mice. Previous studies had demonstrated that either synthetic or naturally
Shinji Oikawa et al.
Free radical research, 40(1), 31-39 (2005-11-22)
4-Hydrazinobenzoic acid, an ingredient of mushroom Agaricus bisporus, is carcinogenic to rodents. To clarify the mechanism of carcinogenesis, we investigated DNA damage by 4-hydrazinobenzoic acid using (32)P-labeled DNA fragments obtained from the human p53 and p16 tumor suppressor genes. 4-Hydrazinobenzoic
K Walton et al.
Mutation research, 381(1), 131-139 (1997-12-24)
Agaritine (N2-[L-(+)-glutamyl]-4-(hydroxymethylphenyl)hydrazine), the principal hydrazine found in the edible mushroom Agaricus bisporus, as well as the N'-acetyl derivative of 4-(hydroxymethyl)phenylhydrazine and 4-(hydroxymethyl)benzene diazonium ion, as the tetraborate salt, considered as the putative proximate and ultimate carcinogens of agaritine, were all

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service