Skip to Content
Merck
All Photos(2)

Key Documents

432431

Sigma-Aldrich

Polystyrene-block-poly(ethylene-ran-butylene)-block-polystyrene-graft-maleic anhydride

melt index ~21 g/10 min (230°C/5.0kg)

Synonym(s):

maleated copolymer, maleated polymer, maleic copolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

pellets

Quality Level

melt index

~21 g/10 min (230°C/5.0kg)

composition

maleic anhydride, ~2 wt. %

hardness

75 (Shore A)

viscosity

12.5 poise, 15 wt. % in toluene(25 °C, Brookfield)(lit.)

solubility

solvents with solubility parameters between 7.7 and 9.4: soluble

density

0.91 g/mL at 25 °C (lit.)

elongation

ASTM D 412 - 500% (ultimate)

SMILES string

C=C.C=CC=C.O=C1OC(=O)C=C1.C=Cc2ccccc2

InChI key

WBWITWZSMYQPNN-UHFFFAOYSA-N

Application

Component of hot melt adhesives, sealants, asphalt and oil gels. Modifier for polymers, thermosets and general rubber compounding. Direct molded or extruded auto parts, sporting goods, footware and film.

Features and Benefits

Maleic anhydride is grafted on the mid-block to increase compatibility with polar coatings and increase interaction with substrates. More oxidatively stable than unsaturated precursor. Flexible to -80°C.

Physical form

Thermoplastic elastomer.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramon Tejada-Oliveros et al.
Molecules (Basel, Switzerland), 26(1) (2021-01-21)
This research work reports the potential of maleinized linseed oil (MLO) as biobased compatibilizer in polylactide (PLA) and a thermoplastic elastomer, namely, polystyrene-b-(ethylene-ran-butylene)-b-styrene (SEBS) blends (PLA/SEBS), with improved impact strength for the packaging industry. The effects of MLO are compared

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service