Skip to Content
Merck
All Photos(2)

Key Documents

348937

Sigma-Aldrich

Lithium isopropoxide

95%

Synonym(s):

Lithium 2-propanolate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOLi
CAS Number:
Molecular Weight:
66.03
Beilstein:
3618614
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

powder and chunks

reaction suitability

core: lithium

SMILES string

[Li+].CC(C)[O-]

InChI

1S/C3H7O.Li/c1-3(2)4;/h3H,1-2H3;/q-1;+1

InChI key

HAUKUGBTJXWQMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-heat. 1 - Skin Corr. 1B

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Few Monolayer Atomic Layer Deposition (ALD) on Surfaces and Interfaces for Energy Applications

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service