Skip to Content
Merck
All Photos(1)

Key Documents

274089

Sigma-Aldrich

2-Chlorobenzoxazole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4ClNO
CAS Number:
Molecular Weight:
153.57
Beilstein:
115982
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.566 (lit.)

bp

201-202 °C (lit.)

mp

7 °C (lit.)

density

1.345 g/mL at 25 °C (lit.)

SMILES string

Clc1nc2ccccc2o1

InChI

1S/C7H4ClNO/c8-7-9-5-3-1-2-4-6(5)10-7/h1-4H

InChI key

BBVQDWDBTWSGHQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chlorobenzoxazole was used in the synthesis of 2-(2-naphthylamino)benzoxazole via reaction with 2-amino-1-naphthalenesulfonic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction of 2-chlorobenzoxazole with anthranilic acids and 2-amino-1-naphthalenesulfonic acid.
J Sam et al.
Journal of pharmaceutical sciences, 57(8), 1445-1446 (1968-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service