Skip to Content
Merck
All Photos(1)

Key Documents

265160

Sigma-Aldrich

3,4-Difluorobenzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CHO
CAS Number:
Molecular Weight:
142.10
Beilstein:
2241231
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.5 (lit.)

density

1.288 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)c1ccc(F)c(F)c1

InChI

1S/C7H4F2O/c8-6-2-1-5(4-10)3-7(6)9/h1-4H

InChI key

JPHKMYXKNKLNDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,4-Difluorobenzaldehyde was used in the synthesis of 3-benzylidene 20,29-dihydrobetulinic acid derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

156.2 °F - DIN 51758

Flash Point(C)

69 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rama Mukherjee et al.
Bioorganic & medicinal chemistry letters, 14(12), 3169-3172 (2004-05-20)
New 3-O-acyl, 3-benzylidene, 3-hydrazone, 3-hydrazine, 17-carboxyacryloyl ester derivatives of betulinic acid (2-6, 8-11, 13, 17, 18, 21, and 22) were synthesized and evaluated in vitro for anti-angiogenic activity on endothelial cell cytotoxicity, specificity, and tube-formation ability. All derivatives reported here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service