Skip to Content
Merck

175994

Methylamine-d2 deuteriochloride

≥98 atom % D

Synonym(s):

Methylammonium-d3 chloride

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
CH3ND2·DCl
CAS Number:
Molecular Weight:
70.54
UNSPSC Code:
12352202
PubChem Substance ID:
EC Number:
238-847-5
MDL number:
Isotopic purity:
≥98 atom % D
Mass shift:
M+3
Form:
solid


isotopic purity

≥98 atom % D

Quality Segment

form

solid

mp

233-235 °C (dec.) (lit.)

mass shift

M+3

SMILES string

Cl[2H].[2H]N([2H])C

InChI

1S/CH5N.ClH/c1-2;/h2H2,1H3;1H/i/hD3

InChI key

NQMRYBIKMRVZLB-ZRLBSURWSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library