175773
Cyclohexanol-d12
≥98 atom % D, ≥98% (CP)
Synonym(s):
Cyclohexyl alcohol-d12
About This Item
Recommended Products
vapor pressure
1 mmHg ( 21 °C)
isotopic purity
≥98 atom % D
Assay
≥98% (CP)
autoignition temp.
572 °F
expl. lim.
12.25 %
refractive index
n20/D 1.4608 (lit.)
bp
160-161 °C (lit.)
mp
20-22 °C (lit.)
density
1.059 g/mL at 25 °C
mass shift
M+12
SMILES string
[2H]OC1([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]
InChI
1S/C6H12O/c7-6-4-2-1-3-5-6/h6-7H,1-5H2/i1D2,2D2,3D2,4D2,5D2,6D,7D
InChI key
HPXRVTGHNJAIIH-BFHGLDALSA-N
Related Categories
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
154.4 °F - closed cup
Flash Point(C)
68 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service