Skip to Content
Merck
All Photos(1)

Key Documents

153494

Sigma-Aldrich

2-Amino-5-nitrobenzonitrile

95%

Synonym(s):

2-Cyano-4-nitroaniline, 5-Nitroanthranilonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3(NH2)CN
CAS Number:
Molecular Weight:
163.13
Beilstein:
1425714
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

mp

200-207 °C (lit.)

SMILES string

Nc1ccc(cc1C#N)[N+]([O-])=O

InChI

1S/C7H5N3O2/c8-4-5-3-6(10(11)12)1-2-7(5)9/h1-3H,9H2

InChI key

MGCGMYPNXAFGFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syed Muhammad Saad et al.
European journal of medicinal chemistry, 108, 13-20 (2015-12-01)
4-Arylamino-6-nitroquinazolines (2-25) were synthesized and evaluated for their leishmanicidal activities against Leishmania major promastigotes in vitro with IC50 values = 1.87-61.48 μM. Among the twenty four synthetic derivatives, 4-[4'-(methylsulfanyl)phenyl]amino-6-nitroquinazoline (21), and 4-(2'-methoxyphenyl)amino-6-nitroquinazoline (8) showed excellent antileishmanial activities with IC50 values 1.87 ± 0.31 and 4.37 ± 0.02 μM, respectively
Alexis Enright et al.
The Analyst, 129(10), 975-978 (2004-10-01)
A series of eleven specially designed benzotriazole monoazo dyes for use in surface enhanced resonance Raman scattering studies are reported. Unlike previous benzotriazole dyes produced for SERRS, these dyes have been synthesised to be trifunctional in nature. The presence of
Jinho Lee et al.
Bioorganic & medicinal chemistry letters, 18(7), 2292-2295 (2008-03-21)
A novel series of 3,5-diaminoindazoles were prepared and found to be CDK inhibitors. Potent inhibitors against CDK1 and CDK2 were obtained by introduction of 1lambda(6)-isothiazolidine-1,1-dioxide at 5-position of indazole. Anti-proliferative activities of compounds were evaluated using EJ, HCT116, SW620, and
Anton Georgiev et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 175, 76-91 (2016-12-27)
In this paper three different "push-pull" 4-aminoazobenzene dyes have been synthesized in order to characterize their photochromic behavior in different solvents. The molecular geometry was optimized by DFT/B3LYP functional combined with the standard 6-31+G(d,p) basis set for trans (E) and
Jie Hu et al.
Chemical biology & drug design, 85(6), 672-684 (2014-10-21)
Quinazoline has been reported to exhibit multiple bioactivities. The aim of this study was to discover new quinazoline derivatives with preventive effect on lipopolysaccharide-induced acute lung injury via anti-inflammatory actions. Thirty-three 4-amino quinazolin derivatives were synthesized and screened for anti-inflammatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service