Skip to Content
Merck
All Photos(1)

Key Documents

129488

Sigma-Aldrich

5-Iodovanillin

97%

Synonym(s):

4-Hydroxy-3-iodo-5-methoxybenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H2(OCH3)(OH)CHO
CAS Number:
Molecular Weight:
278.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

powder

mp

183-185 °C (lit.)

functional group

aldehyde
iodo

SMILES string

COc1cc(C=O)cc(I)c1O

InChI

1S/C8H7IO3/c1-12-7-3-5(4-10)2-6(9)8(7)11/h2-4,11H,1H3

InChI key

FBBCSYADXYILEH-UHFFFAOYSA-N

Application

5-Lodovanillin was used in the characterization of 5-Iodoacetovanillone[1]. 5-Iodovanillin has also been used in the synthesis of platelet activating factor antagonist L-659,989[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

An improved synthesis of acetosyringone.
Crawford LW, et al.
Canadian Journal of Chemistry, 34(11), 1562-1566 (1956)
Thompson AS, et al.
Tetrahedron Letters, 31(48), 6953-6956 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service