Merck
  • α-Bromo-2,6-dichlorotoluene: molecular structure, vibrational spectroscopy, natural bond orbital analysis and NMR studies.
α-Bromo-2,6-dichlorotoluene: molecular structure, vibrational spectroscopy, natural bond orbital analysis and NMR studies.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-05-23)
V Karunakaran, V Balachandran
ABSTRACT
The FT-IR and FT-Raman spectra of α-bromo-2,6-dichlorotoluene (αBDCT) have been recorded. The structural and spectroscopic data of the molecule in the ground state have been calculated using Hartree Fock (HF) and Density Functional Theory (DFT)/B3LYP with the standard 6-31++G(d,p) basis set. The optimized molecular geometry, vibrational frequencies and atomic charges in the ground state are calculated. With the help of specific scaling procedures, the observed vibrational wavenumbers in FT-IR and FT-Raman spectra have been analyzed and assigned to different normal modes of the molecule. A detailed interpretation of the infrared and Raman spectra of αBDCT is also reported based on total energy distribution (TED). The (1)H and (13)C NMR chemical shifts have been calculated by gauge-including atomic orbital method with B3LYP/6-31++G(d,p) approach. Stability of the molecule arising from hyperconjugative interactions and charge delocalization has been analyzed using natural bond orbital (NBO) analysis. The theoretical results agree well with the observed spectra.
Sigma-Aldrich
2,4-Dichlorotoluene, 99%