Key Documents
Z227250
Supelco® PEEK Fitting
One-piece fingertight, pkg of 5 ea
Synonym(s):
PEEK Fitting
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
Pricing and availability is not currently available.
packaging
pkg of 5 ea
General description
Biocompatible fittings for all 1/16 in. tubing connections.
Features and Benefits
Holds to 6000 psi (420 kg/cm2).
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1