LNEBIO101L1
Glucose, Creatinine, Glycerides, Cholesterol, in human frozen serum
LNE Certified Reference Material, commutable, JCTLM listed
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352211
NACRES:
NA.24
Recommended Products
form
liquid
quality
commutable, JCTLM listed
shelf life
limited shelf life, expiry date on the label
composition
human serum, 100%
IVD
not for in vitro diagnostic use
storage temp.
−70°C
Looking for similar products? Visit Product Comparison Guide
General description
LNE CRM Bio 101L1 is a JCTLM database listed, higher-order certified reference material produced by the French metrological institute LNE in compliance with ISO 17034 requirements.
The Certified Reference Materials LNE CRM Bio 101L1 and LNE CRM Bio 101L2 correspond to 2 pools of Human Frozen Serum that were prepared according to the Clinical Laboratory Standards Institute (CLSI) C37-A guidelines describing the preparation and validation of commutable frozen human serum pools as Secondary Reference Materials for Cholesterol Measurement. Samples commutability was assessed through 2 commutability studies according to the difference in bias approach described in the IFCC recommendations on commutability assessment involving several clinical laboratories. Commutability results are reported in the certificate of analysis.
Cert_ LNE CRM Bio 101L1
The Certified Reference Materials LNE CRM Bio 101L1 and LNE CRM Bio 101L2 correspond to 2 pools of Human Frozen Serum that were prepared according to the Clinical Laboratory Standards Institute (CLSI) C37-A guidelines describing the preparation and validation of commutable frozen human serum pools as Secondary Reference Materials for Cholesterol Measurement. Samples commutability was assessed through 2 commutability studies according to the difference in bias approach described in the IFCC recommendations on commutability assessment involving several clinical laboratories. Commutability results are reported in the certificate of analysis.
Cert_ LNE CRM Bio 101L1
Application
LNE CRM Bio 101L1, a secondary standard, is intended for use as quality control material to assess the measurement bias or measurement uncertainty of established or new measurement procedures for the determination of creatinine, glucose, total cholesterol, total glycerides, HDL-cholesterol and LDL-cholesterol in human serum.
Features and Benefits
- A reliable Certified Reference Material (CRM) for clinically relevant biomarkers
- Concentrations of glucose, creatinine, total cholesterol, triglyceride (TG), high-density lipoprotein (HDL)-cholesterol and low-density lipoprotein (LDL)-cholesterol are certified by higher-order reference methods.
- Derived from real patient samples
- This CRM is pooled, tested, and certified by the LNE, which includes extensive commutability studies.
- The accuracy of the reference methods used to certify target values is verified using certified matrix reference materials.
- Listed by the JCTLM as higher-order reference materials.
Other Notes
- Details on storage, safety, usage, and operating instructions are provided in the comprehensive Certificate of Analysis (CoA).
- Concentrations of glucose, creatinine, total glycerides and total cholesterol are certified by reference measurement procedures relying on Gas Chromatography Isotope Dilution mass Spectrometry (GC-IDMS) listed in JCTLM database.
- Concentrations of HDL-C and LDL-C are certified at the CDC by Beta Quantification with cholesterol being determined by the Abell Kendall method.
- This clinical matrix CRM is for research and clinical testing applications.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service