G005396
HPLC Analysis of Chinese Ginseng Extract on Ascentis® Express C18
application for HPLC
About This Item
Recommended Products
technique(s)
HPLC: suitable
test parameters
column: Ascentis Express C18, 15 cm x 4.6 mm I.D., 2.7 μm particles (53829-U)
mobile phase: [A] water; [B] acetonitrile
gradient: held at 25% B for 1.5 min; 25 to 85% B in 12 min; held at 85% for 1 min
flow rate: 1 mL/min
pressure: 3481 psi (240 bar)
column temp.: 60 °C
detector: UV, 205 nm
injection: 25 μL
sample: Extracted from dried, ground plant material; 10 mg/mL in water:acetonitrile (85:15); sonicate 10 min at ambient
suitability
application for HPLC
application(s)
vitamins, nutraceuticals, and natural products
Analysis Note
Legal Information
Analyte
analytical column
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service