G004415
Improved HPLC Selectivity of Discovery® BIO Wide Pore RP Phases
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
column: Discovery BIO Wide Pore C18, 15 cm x 4.6 mm I.D., 5 μm particles vs. competitive wide pore C18 column of same dimensions (568222-U)
mobile phase: [A] 0.1% TFA in 80:20 water:acetonitrile; [B] 0.1% TFA in 66:34 water:acetonitrile
Gradient of 0 to 100% B in 14 min. after 1 min. delay
gradient: 0 to 100% B in 14 min. after 1 min. delay
flow rate: 1 mL/min
column temp.: 30 °C
detector: UV, 220 nm
injection: 10 μL
sample: peptide mixture (Sigma P2693), ~0.25 μg each peptide in 0.1% TFA
suitability
application for HPLC
application(s)
life science and biopharma
Legal Information
analytical column
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service