Skip to Content
Merck
All Photos(1)

Key Documents

814686

TOYOPEARL® HW-55S Bulk Media

phase hydroxylated methacrylic polymer, bottle of 1 L, 30 μm particle size

Synonym(s):

TOYOPEARL® HW-55 Size Exclusion Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
particle size:
30 μm
matrix active group:
hydroxylated methacrylic polymer phase
matrix:
hydroxylated methacrylic polymer

product line

TOYOPEARL®

form

slurry

packaging

bottle of 1 L

manufacturer/tradename

Tosoh 14686

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

hydroxylated methacrylic polymer phase

particle size

30 μm

operating pH

2-13

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl HW-55 resin is a hydroxylated methacrylic polymer bead for size exclusion chromatography of proteins between 1,000-700,000 Da. It also is the base bead for all Toyopearl "-550" designated products.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan Rui et al.
ACS applied materials & interfaces, 11(11), 10472-10480 (2019-02-23)
Functional codelivery of plasmid DNA and RNA oligonucleotides in the same nanoparticle system is challenging due to differences in their physical properties as well as their intracellular locations of function. In this study, we synthesized a series of reducible branched

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service