Skip to Content
Merck
All Photos(1)

Key Documents

67274-U

Supelco

BIOshell A160 Peptide C18 (2 µm) HPLC Columns

L × I.D. 7.5 cm × 3.0 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.52
L × I.D.:
7.5 cm × 3.0 mm
surface area:
65 m2/g
particle size:
2.0 μm
matrix active group:
C18 (octadecyl) phase
pore size:
160 Å pore size
matrix:
spherical silica particle platform
superficially porous particle

Product Name

BIOshell A160 Peptide C18 Column, 2 µm HPLC Column, 2.0 μm particle size, L × I.D. 7.5 cm × 3.0 mm

material

stainless steel hardware

Agency

suitable for USP L1

product line

BIOshell

feature

endcapped: no

packaging

1 ea of

parameter

14,500 psi max. pressure (1000 bar)
60 °C max. temp.

technique(s)

HPLC: suitable
LC/MS: suitable
UHPLC-MS: suitable
UHPLC: suitable

L × I.D.

7.5 cm × 3.0 mm

surface area

65 m2/g

matrix

spherical silica particle platform
superficially porous particle

matrix active group

C18 (octadecyl) phase

particle size

2.0 μm

pore size

160 Å pore size

operating pH range

1-8

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Shell thickness: 0.4 μm
Solid Core: 1.2 μm
Extent of labeling: 4.0%

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service