Skip to Content
Merck
All Photos(1)

Key Documents

61092

Supelco

TraceCERT® Multiresidue Pesticide Standard Mix #10

in acetonitrile, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

in acetonitrile
100 μg/mL each component in acetonitrile

application(s)

cannabis testing
food and beverages
(pesticide residue testing)

format

multi-component solution

storage temp.

−20°C

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Other Notes

18 components. When combined with other mixes, suitable for use in meeting the requirements of mulitple jurisdictions for pesticide testing of cannabis products
Check out our entire range of pesticide mixes for Cannabis testing

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Methoprene, reference material
    Benzovindiflupyr, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
    Buprofezin, reference material
    Cyantraniliprole, reference material
    Cyprodinil, reference material
    Deltamethrin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
    Fluopyram, reference material
    Fensulfothion, reference material
    Pyraclostrobin, reference material
    Pirimicarb, reference material
    Spirodiclofen, reference material
    Resmethrin, reference material
    Phenothrin
    Tetrachlorvinphos, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
    Tetramethrin
    Fenthion
    Dodemorph
    Allethrin
    See All (18)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct extraction with acetonitrile of hemp seed oil for the analysis of pesticides by using comprehensive two-dimensional gas chromatography-triple quadrupole mass spectrometry
Arena A, et al.
Journal of Chromatography A, 464432-464432 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service