Key Documents
57048
Supelco
Long Stem Flow Control Valves
pk of 6
About This Item
UNSPSC Code:
12000000
packaging
pk of 6
Quality Level
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
General description
Equip alternative valves in your 12-port or 24-port standard Visiprep vacuum manifold with these long stem flow control valves if you intend to use all ports of the manifold with 12, 20, or 60 mL SPE tubes.
Note: Not for use with Visiprep DL (Disposable Liner) models
Choose from one of the most recent versions:
Certificates of Analysis (COA)