505374
Supelclean™ LC-Si SPE Tube
bed wt. 500 mg, volume 6 mL, pk of 30
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115712
Recommended Products
material
PE frit
polypropylene hardware
product line
Supelclean™
composition
bed wt., 500 mg
packaging
pk of 30
technique(s)
solid phase extraction (SPE): suitable
surface area
475 m2/g
volume
6 mL
matrix
silica gel base material (irregularly shaped)
matrix active group
silica bonding
particle size
45 μm
pore size
0.8 cm3/g pore volume
60 Å pore size
application(s)
food and beverages
separation technique
normal phase
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Supelclean™ LC-Si SPE Tube is used to adsorb moderately polar to polar compounds from nonaqueous samples.
Legal Information
Supelclean is a trademark of Sigma-Aldrich Co. LLC
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Zhihui Feng et al.
Food research international (Ottawa, Ont.), 129, 108841-108841 (2020-02-11)
The orchid-like odor of a tea infusion is regarded as a noble aroma and an essential sensory attribute for certain premium teas. Such tea leaves are difficult to make and the quality is not always reproducible. This study is focused
Zhihui Feng et al.
Food chemistry, 285, 347-354 (2019-02-25)
Tea aroma is determined by the nature of the plant, the production processes, and many other factors influencing its formation and release. The objective of this study was to investigate the impact of manufacturing processes on the aroma composition of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service