ZRB2086
Anti-CD167a/DDR1 Antibody, clone 1D11 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells
Synonym(s):
Epithelial discoidin domain-containing receptor 1;EC:2.7.10.1;Epithelial discoidin domain receptor 1;CD167 antigen-like family member A;Cell adhesion kinase;Discoidin receptor tyrosine kinase;HGK2;Mammary carcinoma kinase
About This Item
Recommended Products
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
1D11, recombinant monoclonal
description
1D11 Clone
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 101.13 kDa
observed mol wt ~95 kDa
purified by
using Protein A
species reactivity
human
species reactivity (predicted by homology)
porcine, mouse, rat, monkey, bovine
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
C-terminal half
Protein ID accession no.
UniProt accession no.
greener alternative category
, Aligned
shipped in
ambient
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... DDR1(780)
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in T47D cell lysate.
Western Blotting Analysis: A 1:10,000 dilution of this antibody detected CD167a/DDR1 in T47D cell lysate.
Tested Applications
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CD167a/DDR1 in human small intestine tissue sections.
Affinity Binding Assay: A representative lot of this antibody bound CD167a/DDR1 peptide with a KD of 3.5 x 10-7 in an affinity binding assay.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CD167a/DDR1 in MCF-7 cells.
Flow Cytometry Analysis: 0.1 µg from a representative lot detected CD167a/DDR1 in one million MCF-7 cells.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service