Skip to Content
Merck
All Photos(9)

Key Documents

ZMS1193

Sigma-Aldrich

Anti-Trop2 Antibody, clone 2G10 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-CD43 Antibody, clone 1O6 ZooMAb® Rabbit Monoclonal, Cell surface glycoprotein Trop-2, Membrane component chromosome 1 surface marker 1, Pancreatic carcinoma marker protein GA733-1, Tumor-associated calcium signal transducer 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2G10, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 35.71 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... TACSTD2(4070)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2G10 is a ZooMAb® mouse recombinant monoclonal antibody that specifically detects Tumor-associated calcium signal transducer 2 (Trop2).

Immunogen

His and hFc-tagged recombinant protein corresponding to the extracellular domain of human Tumor-associated calcium signal transducer 2 (Trop2)

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant fragment of Human Trop2.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected His- and hFc-tagged recombinant fragment of Human Trop2.

Tested Applications

Flow Cytometry Analysis: 1 µg from a representative lot detected Trop2 in one million HaCaT cells.

Enzyme Immunoassay (ELISA) Analysis: A serial of dilutions from a representative lot detected His-and hFc-tagged recombinant fragment of Human Trop2.

Affinity Binding Assay: A representative lot of this antibody bound His and hFc-tagged recombinant fragment of Human Trop2 with a KD of 1.5 x 10-6 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 filution from a representative lot detected Trop2 in HaCaT cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Tumor-associated calcium signal transducer 2 (UniProt: P09758; also known as Cell surface glycoprotein Trop-2, Trophoblast antigen 2, Membrane component chromosome 1 surface marker 1, Pancreatic carcinoma marker protein GA733-1) is encoded by the TACSTD2 (also known as GA733-1, M1S1, TROP2) gene (Gene ID: 4070) in human. Trop2 is a single-pass type 1 membrane glycoprotein that is synthesized with a signal peptide (aa 1-26), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 27-274), a transmembrane domain (aa 275-297), and a cytoplasmic domain (aa 298-323). Its expression is observed in placenta and is highly expressed in many cancers where it plays a role in tumor progression. About 85% of all solid cancers display increased expression of Trop2. Its expression is also reported in stem cells in various tissues. TROP2 expression in cancer cells has been correlated with drug resistance. Upregulation of Trop2 is reported to drive the e

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service