Key Documents
Z757942
Sartorius Picus electronic pipette
8 channel, volume 50-1200 μL, pack of 1 ea
Synonym(s):
electronic pipetting, pipettors
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
AC/DC input
100 V / 240 V AC, European 2-pin plug (flat 2-pin)
100 V / 240 V AC, UK plug (3 pin)
100 V / 240 V AC, US 2-pin plug
feature
8-channel
CE compliant
electronic
packaging
pack of 1 ea
manufacturer/tradename
Sartorius 735391
volume
50-1200 μL
battery power
rechargeable
suitability
suitable for (Product is used in uHPLC, HPLC and trace analysis applications.)
Related Categories
General description
Sartorius Picus of the Biohitfamily is the smallest and lightest electronic pipette on the market helping to avoid pipetting caused symptoms like RSI (repetitive strain injury). The Picus handle design and the location of the operation buttons enables comfortable pipetting.
Electronic tip ejection, one hand-operated adjustment wheel, and conveniently located start button minimizes the force needed in pipetting. Repetitive and time consuming pipetting can be done accurately and fast with serial dispensing modes and a new Tracker - great help for example in microplate work.
  • Tracker in conjunction with pipetting, reverse pipetting and multi dispensing modes to help microplate work
  • Adjustment possibility for 1-point, 2-points and 3-points
  • Easy maintenance of lower parts
  • Safe-Cone filters in all models > 10 μl to prevent contaminations
  • Autoclavable lower parts in all models (excl. mcp 1200 μL)
Choose from one of the most recent versions:
Certificates of Analysis (COA)