Z627410
BRAND® BLAUBRAND® volumetric flask, PP stopper, clear glass
volume 25 mL, accuracy: 0.06 mL, neck joint: ST/NS 12/21, polypropylene stopper
Synonym(s):
Volumetric flask with stopper
About This Item
material
borosilicate glass 3.3 flask
polypropylene stopper
wide-neck
Joint
joint: ST/NS neck12/21
description
calibrated to contain (TC, IN)
feature
class A DIN EN ISO 1042. Calibrated to contain (TC, In).
packaging
pack of 2 ea
manufacturer/tradename
BRAND 37293
volume measuring range
25 mL, accuracy: 0.06 mL
capacity
25 mL
Looking for similar products? Visit Product Comparison Guide
General description
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
Other Notes
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service