Z333905
BRAND® accu-jet® pro pipette controller membrane filters
spare membrane filters 0.2 μm, sterile
Synonym(s):
bottle top filters, laboratory filters, vacuum system filters
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Recommended Products
material
PTFE
sterility
sterile
feature
hydrophobic
packaging
pkg of 1 ea
manufacturer/tradename
BRAND 26530
pore size
0.2 μm pore size
General description
The accu-jet®S / pro / macro pipette controller is designed to safeguard your equipment with its robust PP housing and integrated interior membrane that effectively blocks liquid. This protective feature ensures the longevity of your pipette controller while maintaining a clean and safe working environment.
Other Notes
BR26530 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
accu-jet is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)