Z326879
BRAND® BLAUBRAND® volumetric flask, glass stopper, clear glass
volume 25 mL, accuracy: 0.04 mL, neck joint: ST/NS 10/19, glass stopper
All Photos(1)
About This Item
material
borosilicate glass
glass stopper
Joint
joint: ST/NS neck10/19
sterility
non-sterile
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
closure type Glass stopper
packaging
pack of 2
manufacturer/tradename
BRAND
volume measuring range
25 mL, accuracy: 0.04 mL
capacity
25 mL
General description
Volumetric flasks according to DIN ISO 1042, Class A, conformity certified. Made from Boro 3.3, calibrated to contain (TC,IN). Supplied with a glass stopper. Marks and inscriptions in high contrast blue enamel.
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
Other Notes
BR37258 (India)
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG