WHA77002810
Whatman® UNIFILTER® 96 well filter plates
clear polystyrene, membrane, non-sterile, DNA, 25 ea
Synonym(s):
Z648329, micro plate, multiwell plate, well plates
About This Item
Recommended Products
product name
Whatman® 96 well PCR Clean-up UNIFILTER® filter plate, UNIFILTER, 96 well, 800 μl, DNA binding, clear polystyrene, filter bottom with long drip director, 25/pk
material
clear polystyrene
conical bottom clear wells
filter
description
96 well PCR clean-up filter plate
sterility
non-sterile
packaging
pack of 25
manufacturer/tradename
Whatman 7700-2810
Whatman Article No. 28420812 (US reference)
technique(s)
PCR: suitable
maximum volume
800 μL
size
96 wells
working volume
800 μL
suitability
suitable for (PCR clean-up; purified DNA is ready for sequencing, hybridization assays, and microarrays)
binding type
non-treated surface
Features and Benefits
- No cross talk. Patented integral filter design prevents well-to-well cross contamination.
- Economical to use. Wide range of well volume options ensures efficient use of materials.
- Better control. Choice of filter media allows control of the flow rates and retention characteristics.
- Versatile. A broad range of filtration media is available, including glass fiber, polypropylene, cellulose nitrate, cellulose acetate, nylon, and ion exchange cellulose.
Other Notes
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service