SRP8025
TRAIL, Soluble mouse
recombinant, expressed in E. coli, His tagged, >95% (SDS-PAGE)
Synonym(s):
Apo-2L, CD253, JAG2, TRAIL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
biological source
mouse
recombinant
expressed in E. coli
tag
His tagged
Assay
>95% (SDS-PAGE)
form
liquid
mol wt
~24 kDa
packaging
pkg of 20 μg
concentration
≥0.2 mg/mL
impurities
<0.01 EU/μg endotoxin, tested
color
clear
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
mouse ... Tnfsf10(22035)
General description
Tumor Necrosis Factor (TNF)-Related Apoptosis-Inducing Ligand (TRAIL) is a type II transmembrane protein and belongs to the TNF superfamily of ligands. Trimerisation of TRAIL around a centrally positioned Zn atom is needed for maintaining its stability, solubility and biological activity as a death ligand. TRAIL exists in two forms, soluble and membrane bound.
Biochem/physiol Actions
Tumor Necrosis Factor (TNF)-Related Apoptosis-Inducing Ligand (TRAIL) activates the extrinsic apoptotic pathway by binding death receptors (DR) 4 and 5. In addition to DR 4 and 5, TRAIL activates several other receptors such as decoy receptor (DcR)1, DcR2 and a soluble receptor called osteoprotegerin. TRAIL plays a vital role in the inhibition of autoimmune inflammation and cell cycle progression. TRAIL selectively kills tumor cells without damaging normal tissues and therefore, it might act as a tumor-selective anticancer agent. TRAIL-induced apoptosis is involved in various cellular processes including activation-induced cell death, homeostasis, intrathymic negative selection, autoimmunity suppression, and immune surveillance. Repeated dosage of recombinant soluble TRAIL triggers apoptosis, inhibits tumor progression, and increases viability of mice suffering from cancer.
Physical form
Solution in 20 mM HEPES, pH 7.4, 300 mM NaCl, 0.01% Tween 20, 1% sucrose, 1 mM DTT.
Other Notes
The extracellular domain of mouse TRAIL (aa 99-291) is fused at the N-terminus to a His-tag and a linker peptide. The active multimeric conformation is stabilized by an inserted mutation allowing an additional CC-bridge.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) is an inhibitor of autoimmune inflammation and cell cycle progression.
Song K
The Journal of Experimental Medicine, 191(7), 1095-1104 (2000)
Exosomes secreted by human placenta carry functional Fas ligand and TRAIL molecules and convey apoptosis in activated immune cells, suggesting exosome-mediated immune privilege of the fetus.
Stenqvist AC
Journal of Immunology, 191(11), 5515-5523 (2013)
Tumor necrosis factor (TNF)-related apoptosis-inducing ligand (TRAIL) induced mitochondrial pathway to apoptosis and caspase activation is potentiated by phospholipid scramblase-3.
Ndebele K
Apoptosis, 13(7), 845-856 (2008)
Overexpression of soluble TRAIL induces apoptosis in human lung adenocarcinoma and inhibits growth of tumor xenografts in nude mice.
Shi J
Cancer Research, 65(5), 1687-1692 (2005)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service