Skip to Content
Merck
All Photos(1)

Key Documents

SRP0353

Sigma-Aldrich

Akt1 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥75% (SDS-PAGE)

Synonym(s):

PKB, RAC, v-akt murine thymoma viral oncogene homolog 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥75% (SDS-PAGE)

form

aqueous solution

mol wt

60.2 kDa

packaging

pkg of 10 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... AKT1(207)

General description

AKT1 (v-akt murine thymoma viral oncogene homolog 1) is a Ser/Thr protein kinase, and is also called protein kinase B (PKB). This protein is activated when it is phosphorylated at serine 473 and threonine 308. This gene is localized to human chromosome 14q32.32.

Application

Akt1 active human has been used for the testing of recombinant mouse p21 proteins as its substrates using in vitro protein kinase assay.

Biochem/physiol Actions

AKT1 (v-akt murine thymoma viral oncogene homolog 1) is responsible for the phosphorylation and cytoplasmic localization of CKDN1A (cyclin-dependent kinase (CDK) inhibitor 1, also referred to as p21). However, p21 is one of the CDKNs which normally resides in the nucleus and its cytoplasmic localization, mediated by AKT1, is common to various cancers where it suppresses proteins essential for apoptosis. Studies show that up-regulation of AKT1 serves as a poor prognostic marker in breast cancer, which however, depends upon factors including population, hormone receptor status, and hormonal or trastuzumab therapy status.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vogiatzi P and Giordano A.
Cancer Biology & Therapy, 6 (2007)
Zu-Yao Yang et al.
Scientific reports, 5, 7758-7758 (2015-01-15)
The prognostic value of phosphorylated Akt (pAkt) overexpression in breast cancer has been investigated by many studies with inconsistent results. This systematic review was conducted to evaluate the association of pAkt overexpression with breast cancer prognosis in terms of overall
Coenraad Kuijl et al.
Nature, 450(7170), 725-730 (2007-11-30)
With the emergence of multidrug resistant (MDR) bacteria, it is imperative to develop new intervention strategies. Current antibiotics typically target pathogen rather than host-specific biochemical pathways. Here we have developed kinase inhibitors that prevent intracellular growth of unrelated pathogens such
Edgar Gil Rizzatti et al.
British journal of haematology, 130(4), 516-526 (2005-08-16)
Microarray studies have revealed the differential expression of several genes in mantle cell lymphoma (MCL), but it is unknown which of these differences are dependent on the transformed MCL cell itself or on the tumour microenvironment. To investigate which genes
Christelle de Renty et al.
PloS one, 9(5), e97434-e97434 (2014-05-23)
Proliferating trophoblast stem cells (TSCs) can differentiate into nonproliferating but viable trophoblast giant cells (TGCs) that are resistant to DNA damage induced apoptosis. Differentiation is associated with selective up-regulation of the Cip/Kip cyclin-dependent kinase inhibitors p57 and p21; expression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service