Skip to Content
Merck
All Photos(2)

Key Documents

SRP0122

Sigma-Aldrich

LSD1 Active human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE)

Synonym(s):

Amine oxidase (flavin containing) domain 2 (AOF2), BRAF35-HDAC complex protein BHC110, KIAA0601, Lysine (K)-specific demethylase 1 (LSD1), Lysine-specific histone demethylase 1A (KDM1A)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

aqueous solution

mol wt

103 kDa

packaging

pkg of 50 μg

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KDM1A(23028)

General description

Human LSD1, AOF2, amine oxidase, flavin containing, domain 2, GenBank Accession No. NM_015013, amino acids 158-end with N-terminal ST tag, MW = 103 kDa, expressed in E. coli expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

One unit is defined as the amount of enzyme required to demethylate 1 pmol of substrate/min at 37°C.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20 and 10% glycerol.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lama AlAbdi et al.
Cell reports, 30(5), 1478-1490 (2020-02-06)
An aberrant increase in pluripotency gene (PpG) expression due to enhancer reactivation could induce stemness and enhance the tumorigenicity of cancer stem cells. Silencing of PpG enhancers (PpGe) during embryonic stem cell differentiation involves Lsd1-mediated H3K4me1 demethylation and DNA methylation. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service