Skip to Content
Merck
All Photos(1)

Key Documents

SML3374

Sigma-Aldrich

Eurycomanone

≥95% (HPLC)

Synonym(s):

(1β,11β,12α,15β)-11,20-Epoxy-1,11,12,14,15-pentahydroxypicrasa-3,13(21)-diene-2,16-dione, NSC 339187, Pasakbumin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O9
CAS Number:
Molecular Weight:
408.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O1[C@@]2([C@H]3[C@]4([C@H](OC(=O)[C@@H]([C@]4(C(=C)[C@H]2O)O)O)C[C@@H]5[C@@]3([C@@H](C(=O)C=C5C)O)C)C1)O

InChI

1S/C20H24O9/c1-7-4-10(21)13(23)17(3)9(7)5-11-18-6-28-20(27,16(17)18)12(22)8(2)19(18,26)14(24)15(25)29-11/h4,9,11-14,16,22-24,26-27H,2,5-6H2,1,3H3/t9-,11+,12+,13+,14-,16+,17+,18+,19-,20-/m0/s1

InChI key

UCUWZJWAQQRCOR-OKNZMGBLSA-N

Biochem/physiol Actions

Eurycomanone is a main ingredient of commonly consumed herbal preparation Eurycoma longifolia Jack that exhibits antimalarial, antipyretic, aphrodisiac, and cytotoxic activities. Eurycomanone improves spermatogenesis by inhibition of aromatase conversion of testosterone to estrogen, and phosphodiesterase inhibition.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Norzahirah Ahmad et al.
Pharmaceutics, 10(3) (2018-07-13)
Eurycoma longifolia is one of the commonly consumed herbal preparations and its major chemical compound, eurycomanone, has been described to have antimalarial, antipyretic, aphrodisiac, and cytotoxic activities. Today, the consumption of E. longifolia is popular through the incorporation of its
Putri Ayu Jayusman et al.
Nutrients, 10(7) (2018-06-24)
Male osteoporosis is associated with higher rates of disability and mortality. Hence the search for suitable intervention and treatment to prevent the degeneration of skeletal health in men is necessary. Eurycoma longifolia (EL), a traditional plant with aphrodisiac potential may
Bin-Seng Low et al.
Journal of ethnopharmacology, 149(1), 201-207 (2013-07-03)
Eurycoma longifolia Jack (Simaroubaceae family), known locally as 'Tongkat Ali' by the ethnic population, is popularly taken as a traditional remedy to improve the male libido, sexual prowess and fertility. Presently, many tea, coffee and carbonated beverages, pre-mixed with the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service