Skip to Content
Merck
All Photos(1)

Key Documents

SML2645

Sigma-Aldrich

L-Homocitrulline

≥95% (HPLC)

Synonym(s):

Homocitrulline, N6-(Aminocarbonyl)-L-lysine, NSC 27428

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15N3O3
CAS Number:
Molecular Weight:
189.21
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NCCCC[C@@H](C(O)=O)N)N

InChI

1S/C7H15N3O3/c8-5(6(11)12)3-1-2-4-10-7(9)13/h5H,1-4,8H2,(H,11,12)(H3,9,10,13)/t5-/m0/s1

InChI key

XIGSAGMEBXLVJJ-YFKPBYRVSA-N

Biochem/physiol Actions

L-Homocitrulline is a natural amino acid amino acid found in milk of mammalian.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaetano Corso et al.
Dementia and geriatric cognitive disorders extra, 7(1), 143-159 (2017-06-20)
Abnormalities in the plasma amino acid profile have been reported in Alzheimer disease (AD), but no data exist for the prodromal phase characterized by subjective memory complaint (SMC). It was our aim to understand if serum amino acid levels change
Sigurd Delanghe et al.
Nature reviews. Nephrology, 13(9), 580-593 (2017-08-02)
Protein carbamoylation is a non-enzymatic post-translational modification that binds isocyanic acid, which can be derived from the dissociation of urea or from the myeloperoxidase-mediated catabolism of thiocyanate, to the free amino groups of a multitude of proteins. Although the term
Sander De Bruyne et al.
Toxins, 12(2) (2020-01-30)
Carbamoylation is an important risk factor for accelerated atherogenesis and mortality in patients undergoing hemodialysis (HD). We intended to explore whether carbamoylation as assessed by near-infrared (NIR) analysis of nail proteins is associated with (a) serum concentrations of representative uremic
Michael S Lustgarten et al.
Experimental gerontology, 100, 1-10 (2017-10-17)
Reduced skeletal muscle density in older adults is associated with insulin resistance, decreased physical function, and an increased all-cause mortality risk. To elucidate mechanisms that may underlie the maintenance of skeletal muscle density, we conducted a secondary analysis of previously

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service