Skip to Content
Merck
All Photos(1)

Documents

SML2492

Sigma-Aldrich

Engeletin

≥98% (HPLC)

Synonym(s):

(+)-(2R,3R)-Dihydrokaempferol 3-O-α-L-rhamnoside, (2R,3R)-3-[(6-Deoxy-α-L-mannopyranosyl)oxy]-2,3-dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, (2R,3R)-Engelitin, Dihydrokaempferol 3-O-α-L-rhamnopyranoside, Dihydrokaempferol 3-rhamnoside, Engelitin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22O10
CAS Number:
Molecular Weight:
434.39
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -13 to -17°, c = 0.5 in pyridine

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

InChI

1S/C21H22O10/c1-8-15(25)17(27)18(28)21(29-8)31-20-16(26)14-12(24)6-11(23)7-13(14)30-19(20)9-2-4-10(22)5-3-9/h2-8,15,17-25,27-28H,1H3/t8-,15-,17+,18+,19+,20-,21-/m0/s1

InChI key

VQUPQWGKORWZII-WDPYGAQVSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

New MCE product August 2018. Engeletin is a flavanonol glycoside present in the skin of white grapes and white wine that exhibits anti-inflammatory activities. It inhibits LPS induced cytokines production in mouse J774A.1 macrophages. Also, engeletin attenuates LPS-induced endometritis in mice through inhibiting NF-κB activation, and inhibits inflammatory response in LPS-induced acute lung injury (ALI).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xian Jiang et al.
Inflammation, 41(4), 1259-1265 (2018-04-29)
Acute lung injury (ALI) is a serious disease with morbidity and mortality in patients. Engeletin (dihydrokaempferol 3-rhamnoside) is a flavanonol glycoside. It can be found in the skin of white grapes and white wine and is widely distributed in southeast
Srisopa Ruangnoo et al.
Asian Pacific journal of allergy and immunology, 30(4), 268-274 (2013-02-12)
Smilax corbularia is a Thai medicinal plant locally known as 'Hua-Khao-Yen Neua', which is used for treating inflammatory conditions. To evaluate the anti-inflammatory effect of S. corbularia extracts and its isolated compounds by determination of inhibitory effects on lipopolysaccharide-stimulated PGE2
Haichong Wu et al.
Journal of agricultural and food chemistry, 64(31), 6171-6178 (2016-07-15)
Engeletin (dihydrokaempferol 3-rhamnoside) is a flavanonol glycoside. It can be found in the skin of white grapes and white wine and is widely distributed in southeast Asia, and the leaves are used in a tea. Here, we explored the impact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service