Skip to Content
Merck
All Photos(1)

Key Documents

SML2027

Sigma-Aldrich

L-2-oxothiazolidine-4-carboxylic acid propargyl amide

≥98% (HPLC)

Synonym(s):

(R)-2-Oxo-N-2-propyn-1-yl-4-thiazolidinecarboxamide, OTC propargyl amide, OTC-PA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N2O2S
CAS Number:
Molecular Weight:
184.22
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

C#CCNC([C@@H]1CSC(N1)=O)=O

Biochem/physiol Actions

L-2-oxothiazolidine-4-carboxylic acid propargyl amide is a cell penetrant, potent and selective competitive inhibitor of cystathionine-γ-lyase that completely abrogated L-cysteine-induced vasorelaxation in tissue.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service