Skip to Content
Merck
All Photos(1)

Key Documents

SML1874

Sigma-Aldrich

DM-NOFD

≥98% (HPLC)

Synonym(s):

Dimethyl N-oxalyl-D-phenylalanine, N-(2-Methoxy-2-oxoacetyl)-D-phenylalanine methyl ester, N-Oxalyl-D-phenylalanine dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15NO5
CAS Number:
Molecular Weight:
265.26
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

oil

color

, colorless to dark yellow

storage temp.

2-8°C

SMILES string

O=C(C(N[C@H](CC1=CC=CC=C1)C(OC)=O)=O)OC

InChI

1S/C13H15NO5/c1-18-12(16)10(14-11(15)13(17)19-2)8-9-6-4-3-5-7-9/h3-7,10H,8H2,1-2H3,(H,14,15)/t10-/m1/s1

InChI key

CAUUPQLNYYFMOQ-SNVBAGLBSA-N

General description

Dimethyl N-oxalyl-D-phenylalanine (DM-NOFD) is a small FIH (factor-inhibiting) inhibitor molecule.

Biochem/physiol Actions

DM-NOFD is a cell penetrant, prodrug of NOFD a potent and selective inhibitor of an asparaginyl hydroxylase FIH (factor-inhibiting HIF).
DM-NOFD is a cell penetrant, prodrug of NOFD a potent and selective inhibitor of an asparaginyl hydroxylase FIH (factor-inhibiting HIF). DM-NOFD efficiently reduces HIF1a CAD (C-terminal activating domain) hydroxylation in MCF-7 cells.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tuning the transcriptional response to hypoxia by inhibiting HIF prolyl-and asparaginyl-hydroxylases.
Chan MC, et al.
The Journal of Biological Chemistry, jbc-M116 (2016)
Michael A McDonough et al.
Journal of the American Chemical Society, 127(21), 7680-7681 (2005-05-26)
A set of four non-heme iron(II) and 2-oxoglutarate-dependent enzymes catalyze the post-translational modification of a transcription factor, hypoxia inducible factor (HIF), that mediates the hypoxic response in animals. Hydroxylation of HIF both causes its degradation and limits its activity. We
Mun Chiang Chan et al.
The Journal of biological chemistry, 291(39), 20661-20673 (2016-08-10)
The hypoxia-inducible factor (HIF) system orchestrates cellular responses to hypoxia in animals. HIF is an α/β-heterodimeric transcription factor that regulates the expression of hundreds of genes in a tissue context-dependent manner. The major hypoxia-sensing component of the HIF system involves

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service