Skip to Content
Merck
All Photos(1)

Key Documents

SML0317

Sigma-Aldrich

Aldi-2

≥95% (HPLC)

Synonym(s):

3-(Dimethylamino)-1-(3-fluoro-4-methoxyphenyl)-1-propanone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16FNO2
CAS Number:
Molecular Weight:
225.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: >5 mg/mL

storage temp.

2-8°C

SMILES string

COc1ccc(cc1F)C(=O)CCN(C)C

InChI

1S/C12H16FNO2/c1-14(2)7-6-11(15)9-4-5-12(16-3)10(13)8-9/h4-5,8H,6-7H2,1-3H3

InChI key

RNKMLXKYVDDHBX-UHFFFAOYSA-N

Biochem/physiol Actions

Aldi-2 is a potent and specific covalent inhibitor of aldehyde dehydrogenases. Apparently, Aldi-2 undergoes an enzyme-mediated β-elimination which generates a vinyl-ketone intermediate that covalently modifies the active site cysteine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service