Skip to Content
Merck
All Photos(1)

Key Documents

SAB5600193

Sigma-Aldrich

Anti-HA-Tag-Biotin antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM305, purified immunoglobulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

recombinant

expressed in HEK 293 cells

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM305, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

concentration

1 mg/mL

technique(s)

flow cytometry: 0.1-1.0 μg/mL
immunofluorescence: 0.1-1.0 μg/mL
immunohistochemistry: 0.01-0.5 μg/mL
western blot: 0.1-1.0 μg/mL

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

Specificity

This antibody reacts to recombinant proteins containing a HA-Tag fused to either the amino or carboxy terminus. No cross reactivity with other endogenous protein.

Immunogen

A peptide corresponding to HA-tag (Human influenza hemagglutinin amino acids 98-106)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 50% glycerol, 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Igor Patrick Vasconcelos Vieira et al.
AMB Express, 13(1), 131-131 (2023-11-22)
The methylotrophic yeast Komagataella phaffii is one of the most important microbial platforms to produce recombinant proteins. Despite its importance in the context of industrial biotechnology, the use of synthetic biology approaches in K. phaffii is hampered by the fact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service