Skip to Content
Merck
All Photos(6)

Key Documents

SAB1406400

Sigma-Aldrich

Anti-RPA2 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

REPA2, RPA32

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~29.81 kDa

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPA2(6118)

General description

Replication protein A2 (RPA2) is a 32kDa protein. The gene encoding it is localized on human chromosome 1p35. Mouse polyclonal antibody raised against a full-length human RPA2 protein.

Immunogen

RPA2 (AAH12157, 1 a.a. ~ 270 a.a) full-length human protein.

Sequence
MWNSGFESYGSSSYGGAGGYTQSPGGFGSPAPSQAEKKSRARAQHIVPCTISQLLSATLVDEVFRIGNVEISQVTIVGIIRHAEKAPTNIVYKIDDMTAAPMDVRQWVDTDDTSSENTVVPPETYVKVAGHLRSFQNKKSLVAFKIMPLEDMNEFTTHILEVINAHMVLSKANSQPSAGRAPISNPGMSEAGNFGGNSFMPANGLTVAQNQVLNLIKACPRPEGLNFQDLKNQLKHMSVSSIKQAVDFLSNEGHIYSTVDDDHFKSTDAE

Biochem/physiol Actions

Replication protein A2 (RPA2) is involved in DNA repair and replication. The protein has been shown to be phosphorylated as a response to DNA damage and apoptosis in vitro.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhen Zhang et al.
Nucleic acids research, 51(5), 2238-2256 (2023-02-17)
The 53BP1-dependent end-joining pathway plays a critical role in double-strand break (DSB) repair. However, the regulators of 53BP1 in chromatin remain incompletely characterized. In this study, we identified HDGFRP3 (hepatoma-derived growth factor related protein 3) as a 53BP1-interacting protein. The
Proteomic analysis of BRCA1-depleted cell line reveals a putative role for replication protein A2 up-regulation in BRCA1 breast tumor development.
Bouley J
Proteomics. Clinical Applications, 4(5), 489-498 (2010)
Loss of heterozygosity at chromosome 1p in different solid human tumours: association with survival.
Ragnarsson G
British Journal of Cancer, 79(9-10), 1468-1474 (1999)
Phosphorylation of replication protein A middle subunit (RPA32) leads to a disassembly of the RPA heterotrimer.
Treuner K
The Journal of Biological Chemistry, 274(22), 15556-15561 (1999)
Holger Stephan et al.
Nucleic acids research, 37(18), 6028-6041 (2009-08-13)
The human single-stranded DNA-binding protein, replication protein A (RPA), is regulated by the N-terminal phosphorylation of its 32-kDa subunit, RPA2. RPA2 is hyperphosphorylated in response to various DNA-damaging agents and also phosphorylated in a cell-cycle-dependent manner during S- and M-phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service