Skip to Content
Merck
All Photos(3)

Key Documents

PLA0181

Sigma-Aldrich

Rabbit anti-PSF/SFPQ Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

100 kDa DNA-pairing protein, 100 kDa subunit, DNA-binding p52/p100 complex, POMP100, PPP1R140, PSF, PTB-associated splicing factor, PTB-associated-splicing factor, hPOMp100, polypyrimidine tract binding protein associated, polypyrimidine tract-binding protein-associated splicing factor, polypyrimidine tract-binding protein-associated-splicing factor, protein phosphatase 1, regulatory subunit 140, splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated), splicing factor proline/glutamine rich (polypyrimidine tract-binding protein-associated), splicing factor proline/glutamine-rich

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:100- 1:500
immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_005057.1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... PSF/SFPQ(6421)

Immunogen

The epitope recognized by PLA0181 maps to a region between residue 225 and 275 of human Polypyrimidine tract-binding protein-associated-splicing factor (Splicing factor, proline- and glutamine-rich) using the numbering given in entry NP_005057.1 (GeneID 6421).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

PSF/SFPQ is a multifunctional protein know to participate in a variety of nuclear processes and may function to link transcription and pre-mRNA processing. PSF/SFPQ bears an RNA recognition motif (RRM) further indicating its role in post-transcriptional processes. PSF has been shown to associate with p54nrb/NonO to modulate androgen receptor-mediated transcription. Additionally, it has recently been show to associate with XRN2 and p54nrb/NonO to facilitate pre-mRNA 3′ processing and transcription termination.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Zhou et al.
mBio, 10(4) (2019-08-01)
Endogenous retroviruses (ERVs) are transposable elements that cause host genome instability and usually play deleterious roles in disease such as tumorigenesis. Recent advances also suggest that this "enemy within" may encode a viral mimic to induce antiviral immune responses through
Claudia Fierro et al.
Nature communications, 14(1), 3795-3795 (2023-06-27)
The transcription factor ΔNp63 regulates epithelial stem cell function and maintains the integrity of stratified epithelial tissues by acting as transcriptional repressor or activator towards a distinct subset of protein-coding genes and microRNAs. However, our knowledge of the functional link

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service