Skip to Content
Merck
All Photos(1)

Key Documents

N156

Sigma-Aldrich

Naltriben methanesulfonate hydrate

≥98% (HPLC), solid

Synonym(s):

NTB hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H25NO4 · CH4O3S · xH2O
CAS Number:
Molecular Weight:
511.59 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

color

off-white

solubility

DMSO: 14 mg/mL at ~60 °C

storage temp.

−20°C

SMILES string

O.CS(O)(=O)=O.Oc1ccc2C[C@H]3N(CC[C@@]45[C@@H](Oc1c24)c6oc7ccccc7c6C[C@@]35O)CC8CC8

InChI

1S/C26H25NO4.CH4O3S.H2O/c28-18-8-7-15-11-20-26(29)12-17-16-3-1-2-4-19(16)30-22(17)24-25(26,21(15)23(18)31-24)9-10-27(20)13-14-5-6-14;1-5(2,3)4;/h1-4,7-8,14,20,24,28-29H,5-6,9-13H2;1H3,(H,2,3,4);1H2/t20-,24+,25+,26-;;/m1../s1

InChI key

VAGWKZSGWSRENH-HIAJFTJMSA-N

Application

Naltriben methanesulfonate hydrate has been used in the assessment of parasite growth and evaluation of growth inhibition. It has also been used to understand the action of deltorphin-II in exhibiting its antimalarial effect.

Biochem/physiol Actions

Naltriben methanesulfonate is a highly selective δ2 opioid receptor antagonist. The relative ability of the delta antagonists benzylidenenaltrexone and naltriben to inhibit DPDPE-stimulated [35S]GTPgammaS binding suggests that the opioid receptor is of the delta-2 subtype. Ligand binding assays demonstrates biphasic binding of the antagonist to the single receptor type. [35S]GTPgammaS binding is also stimulated by [D-Ser2,Leu5,Thr6]enkephalin > deltorphin II = DPDPE = etorphine > levallorphan = diprenorphine = nalorphine = naltrindole.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Profiling molecular factors associated with pyknosis and developmental arrest induced by an opioid receptor antagonist and dihydroartemisinin in Plasmodium falciparum
Asahi H, et al.
Testing, 12(9), e0184874-e0184874 (2017)
A E Takemori et al.
Life sciences, 50(20), 1491-1495 (1992-01-01)
The pharmacological profile of naltrindole (NTI) and three of its analogues, N-methyl-NTI (N-Me-NTI), oxymorphindole (OMI) and naltriben (NTB) were studied in antinociceptive assays. The compounds were found to have agonist activities that appear to be mediated mainly by kappa opioid
Liliana Matos et al.
Orphanet journal of rare diseases, 9, 180-180 (2014-12-11)
Mutations affecting RNA splicing represent more than 20% of the mutant alleles in Sanfilippo syndrome type C, a rare lysosomal storage disorder that causes severe neurodegeneration. Many of these mutations are localized in the conserved donor or acceptor splice sites
Inhibitory effect of deltorphin-II on development of malaria in Plasmodium berghei-infected mice
Ajitbhai GA, et al.
MalariaWorld journal, 6, 3-3 (2017)
Carla Martins et al.
Human mutation, 40(8), 1084-1100 (2019-06-23)
Mucopolysaccharidosis type IIIC (MPSIIIC) is a severe, rare autosomal recessive disorder caused by variants in the heparan-α-glucosaminide N-acetyltransferase (HGSNAT) gene which result in lysosomal accumulation of heparan sulfate. We analyzed clinical presentation, molecular defects and their haplotype context in 78

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service