Skip to Content
Merck
All Photos(3)

Key Documents

HPA012082

Sigma-Aldrich

Anti-USP36 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Deubiquitinating enzyme 36, Anti-Ubiquitin carboxyl-terminal hydrolase 36, Anti-Ubiquitin thioesterase 36, Anti-Ubiquitin-specific-processing protease 36

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
conjugate:
unconjugated
application:
IF
IHC
clone:
polyclonal
species reactivity:
human
citations:
5
technique(s):
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

DSLVHSSNVKVVLNQQAYVLFYLRIPGSKKSPEGLISRTGSSSLPGRPSVIPDHSKKNIGNGIISSPLTGKRQDSGTMKKPHTTEEIGVPISRNGSTLGLKSQNGCIPPKLPSGSPSPKLSQTPTHMPTILDDPGKKVKKPAPP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... USP36(57602)

General description

USP36 (Ubiquitin specific peptidase 36) is a novel c-Myc deubiquitinase belonging to the ubiquitin-specific protease (USP) family. It is expressed in various human tissues. It is expressed at high levels in skeletal muscle and pancreas and at low levels in heart, placenta, lung, liver, and kidney. This ~61.2kDa protein consists of highly conserved Cys, Asp, His, and Asn/Asp (II) domains.

Immunogen

Ubiquitin carboxyl-terminal hydrolase 36 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

USP36 (Ubiquitin specific peptidase 36) is mainly involved in the nucleolar activities in respect to maintenance of its structural integrity and functions. It cleaves ubiquitin from an ubiquitin-β-galactosidase fusion protein through the deubiquitination process. It also deubiquitylates nucleolar proteins such as nucleophosmin/B23, fibrillarin followed by the stabilization of counteracting ubiquitylation-mediated proteasomal degradation. Studies show its overexpression in several cancerous cells such as human breast and lung cancers.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70776

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Myung-Sun Kim et al.
International journal of oncology, 25(2), 373-379 (2004-07-16)
The regulation of ubiquitin-mediated protein degradation is becoming important for a number of cellular processes. Human HeLa DUB-1 cDNA, encoding a novel deubiquitinating enzyme, was isolated from ovarian cancer cells. It has 1,647 bp nucleotides and encodes a 548 amino
Akinori Endo et al.
Journal of cell science, 122(Pt 5), 678-686 (2009-02-12)
The nucleolus is a subnuclear compartment and the site of ribosome biogenesis. Previous studies have implicated protein ubiquitylation in nucleolar activity. Here we show that USP36, a deubiquitylating enzyme of unknown function, regulates nucleolar activity in mammalian cells. USP36 localized
Xiao-Xin Sun et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(12), 3734-3739 (2015-03-17)
c-Myc protein stability and activity are tightly regulated by the ubiquitin-proteasome system. Aberrant stabilization of c-Myc contributes to many human cancers. c-Myc is ubiquitinated by SCF(Fbw7) (a SKP1-cullin-1-F-box complex that contains the F-box and WD repeat domain-containing 7, Fbw7, as
Víctor Quesada et al.
Biochemical and biophysical research communications, 314(1), 54-62 (2004-01-13)
We have identified and cloned 22 human cDNAs encoding novel members of the ubiquitin-specific protease (USP) family. Eighteen of the identified proteins contain all structural features characteristic of these cysteine proteinases, whereas four of them have been classified as non-peptidase
Jasmin van den Heuvel et al.
eLife, 10 (2021-07-29)
In humans and other holozoan organisms, the ribosomal protein eS30 is synthesized as a fusion protein with the ubiquitin-like protein FUBI. However, FUBI is not part of the mature 40S ribosomal subunit and cleaved off by an as-of-yet unidentified protease.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service