GE29401317
StrepTrap™ XT prepacked chromatography column
pkg of 1 × 1 mL, prepacked chromatography column
Synonym(s):
StrepTrap™ High Performance
About This Item
Recommended Products
grade
for chromatography
form
prepacked column
crosslinking
cross-linked
packaging
pkg of 1 × 1 mL
manufacturer/tradename
Cytiva 29401317
storage condition
(20% Ethanol)
parameter
-4 mL/min max. flow rate
technique(s)
liquid chromatography (LC): suitable
matrix
(Rigid highly cross-linked 6% agarose)
matrix active group
Strep-Tactin XT
particle size
(34 µm)
operating pH
6-10
capacity
binding capacity(10 mg protein with Strep-tag II or Twin-Strep-tag/mL of resin.)
suitability
suitable for chromatography
separation technique
affinity
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Highly pure Strep-tag II and Twin-Strep-tag recombinant proteins.
- Fast purifications with simple elution.
- Physiological conditions and mild elution preserve target protein activity.
- Fast and easy regeneration.
- Compatible with a wide range of detergents, denaturants, and other additives.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
Flash Point(F)
55.4 °F
Flash Point(C)
13 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service