GE28-4065-64
dNTP Set (10 mM each A,C,G,T)
Cytiva 28-4065-64, pack of 500 μL (Total of 500 uL containing 10 mM each A,C,G,T)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106305
NACRES:
NA.52
Recommended Products
form
liquid
packaging
pack of 500 μL (Total of 500 uL containing 10 mM each A,C,G,T)
manufacturer/tradename
Cytiva 28-4065-64
concentration
10 mM
color
Colorless
shipped in
dry ice
storage temp.
−20°C
General description
By incorporating recent advances in manufacturing technology, Cytiva now offers illustra™ Solution dNTPs: the highest purity deoxynucleotides available for amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.
Features and Benefits
- Free from DNase, RNase, and nicking enzyme activity
- Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
- Buffer free and ready-to-use solutions in multiple formats
- Functionally tested for long PCR* and sequencing
- Custom formulation, testing, packaging, and labeling are available
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
illustra is a trademark of Cytiva
Storage Class Code
12 - Non Combustible Liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service