GE17-5279-01
Glutathione Sepharose™ High Performance
Cytiva 17-5279-01, pack of 25 mL
Synonym(s):
Glutathione Sepharose Column
About This Item
Recommended Products
ligand
glutathione
packaging
pack of 25 mL
manufacturer/tradename
Cytiva 17-5279-01
matrix
highly cross-linked 6% agarose
avg. part. size
34 μm
cleaning
3-12
working range
3-12
capacity
>7 mg/mL binding capacity(medium)
General description
Glutathione Sepharose™ High Performance is used for the purification of GST-tagged proteins from bacterial lysates and other S-transferases or glutathione-dependent proteins. GST-tagged proteins are eluted under mild, nondenaturing conditions using reduced glutathione. The purification process preserves protein antigenicity and function.
Features and Benefits
- 34 μm bead size for improved efficiency and resolution
- Fast, one-step purification of GST-tagged proteins and other glutathione S-trasferase or glutathione-dependent proteins
- Mild conditions during the purification process preserves protein antigenicity and function
Storage and Stability
Analysis Note
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
This page describes general considerations for purification of GST-tagged proteins using Cytiva products.
GST-tagged protein yield varies based on protein, host cell, and culture conditions, influencing production.
GST-tagged protein yield varies based on protein, host cell, and culture conditions, influencing production.
GST-tagged protein yield varies based on protein, host cell, and culture conditions, influencing production.
Protocols
This page shows how to purify GST-tagged proteins using Glutathione Sepharose from Cytiva.
GSTrap™ affinity columns are specially designed 1 ml and 5 ml HiTrap® columns packed with Glutathione Sepharose High Performance, Glutathione Sepharose 4 Fast Flow, or Glutathione Sepharose 4B media.
This page how to remove GST tags by enzymatic cleavage using Cytiva products.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service